Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-(+)-2-amino-4-methyl-1-pentanol | 7533-40-6 | MFCD00063676 | 99.5% | 117.19 g·mol⁻¹ | C6H15NO | 1 G
SDP

Catalog No. 5000300262
Encompass_Preferred

L-Leucinol is the L-(+)-enantiomer of leucinol ((S)-(+)-2-amino-4-methyl-1-pentanol), an amino alcohol used as a biochemical reagent and reported competitive aminopeptidase inhibitor. It is provided as a high-purity material for in vitro enzymatic assays and as a synthetic building block in medicinal chemistry.

  • Chemical name: (S)-(+)-2-amino-4-methyl-1-pentanol.
  • CAS number: 7533-40-6.
  • Molecular formula: C6H15NO.
  • Molecular weight: 117.19 g·mol⁻¹.
  • High purity suitable for analytical and synthetic use (≈99.5%).
  • Reported as a competitive aminopeptidase inhibitor (experimental use).
  • Supplied in laboratory-scale quantity (1 g) for research applications.

Catalog No. 50-003-00262 Supplier Medchemexpress LLC Supplier No. HYW0158761G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Leucinol is the L-(+)-enantiomer of leucinol ((S)-(+)-2-amino-4-methyl-1-pentanol), an amino alcohol used as a biochemical reagent and reported competitive aminopeptidase inhibitor. It is provided as a high-purity material for in vitro enzymatic assays and as a synthetic building block in medicinal chemistry.

  • Chemical name: (S)-(+)-2-amino-4-methyl-1-pentanol.
  • CAS number: 7533-40-6.
  • Molecular formula: C6H15NO.
  • Molecular weight: 117.19 g·mol⁻¹.
  • High purity suitable for analytical and synthetic use (≈99.5%).
  • Reported as a competitive aminopeptidase inhibitor (experimental use).
  • Supplied in laboratory-scale quantity (1 g) for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.