Learn More
Medchemexpress LLC (S)-(2-Amino-5-methoxy-4-((triisopropylsilyl)oxy)phenyl)(6-(((tert-butyldimethylsilyl)oxy)methyl... | 2305842-47-9 | C30H54N2O4Si2 | 50 MG
SDP
Catalog No. 5000358260
Encompass_Preferred
(S)-(2-Amino-5-methoxy-4-((triisopropylsilyl)oxy)phenyl)(6-(((tert-butyldimethylsilyl)oxy)methyl)-5-azaspiro[2.4]heptan-5-yl)methanone is a synthetic intermediate useful for pharmaceutical synthesis. It is intended for laboratory use only by experienced personnel.
  • Synthetic intermediate for pharmaceutical synthesis
  • Suitable for laboratory chemical applications
  • Stable under recommended storage conditions
  • Store at 4°C, sealed, away from moisture and light
  • For research use only, not for medical applications
Catalog No. 50-003-58260 Supplier Medchemexpress LLC Supplier No. HY4786850MG
(S)-(2-Amino-5-methoxy-4-((triisopropylsilyl)oxy)phenyl)(6-(((tert-butyldimethylsilyl)oxy)methyl)-5-azaspiro[2.4]heptan-5-yl)methanone is a synthetic intermediate useful for pharmaceutical synthesis. It is intended for laboratory use only by experienced personnel.
  • Synthetic intermediate for pharmaceutical synthesis
  • Suitable for laboratory chemical applications
  • Stable under recommended storage conditions
  • Store at 4°C, sealed, away from moisture and light
  • For research use only, not for medical applications