Learn More
Medchemexpress LLC (S)-2-amino-5-(tert-butoxy)-5-oxopentanoic acid | 2419-56-9 | MFCD00038580 | 98.0% | 203.24 g·mol⁻¹ | C9H17NO4 | 25 G
SDP
Catalog No. 5000304115
Encompass_Preferred
(S)-2-Amino-5-(tert-butoxy)-5-oxopentanoic acid is the S-stereoisomer of a glutamic acid derivative used as a research reagent and synthetic building block for peptide and organic synthesis. It is a white solid with chemical formula C9H17NO4 and molecular weight 203.24 g·mol⁻¹, supplied at high purity for laboratory use.
  • High purity (98.0%) suitable for analytical and synthetic applications
  • S-stereochemistry supports enantioselective synthesis and chiral studies
  • Functionally protected carboxyl group facilitates peptide coupling
  • Compatible with common organic solvents used in synthesis
  • Offered in multiple pack sizes for experimental scale flexibility
  • Documentation available including COA and SDS for safe handling
Catalog No. 50-003-04115 Supplier Medchemexpress LLC Supplier No. 25GHYW014742
(S)-2-Amino-5-(tert-butoxy)-5-oxopentanoic acid is the S-stereoisomer of a glutamic acid derivative used as a research reagent and synthetic building block for peptide and organic synthesis. It is a white solid with chemical formula C9H17NO4 and molecular weight 203.24 g·mol⁻¹, supplied at high purity for laboratory use.
  • High purity (98.0%) suitable for analytical and synthetic applications
  • S-stereochemistry supports enantioselective synthesis and chiral studies
  • Functionally protected carboxyl group facilitates peptide coupling
  • Compatible with common organic solvents used in synthesis
  • Offered in multiple pack sizes for experimental scale flexibility
  • Documentation available including COA and SDS for safe handling