Learn More
Medchemexpress LLC (S)-2-Amino-6-(2-(2-azidoacetamido)acetamido)hexanoic acid (TFA) | 99.77% | 400.31 | 25 MG
SDP
Catalog No. 5000363421
Encompass_Preferred
AzGGK TFA is an unnatural amino acid that can be site-specifically incorporated into proteins through genetic-code expansion. It serves as a site-specific probe for ubiquitylation and SUMOylation. As a click chemistry reagent, AzGGK TFA contains an Azide group, enabling it to undergo various cycloaddition reactions.
  • Unnatural amino acid
  • Site-specifically incorporated into proteins via genetic-code expansion
  • Serves as a site-specific probe for ubiquitylation and SUMOylation
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-63421 Supplier Medchemexpress LLC Supplier No. 25MGHY148835A
AzGGK TFA is an unnatural amino acid that can be site-specifically incorporated into proteins through genetic-code expansion. It serves as a site-specific probe for ubiquitylation and SUMOylation. As a click chemistry reagent, AzGGK TFA contains an Azide group, enabling it to undergo various cycloaddition reactions.
  • Unnatural amino acid
  • Site-specifically incorporated into proteins via genetic-code expansion
  • Serves as a site-specific probe for ubiquitylation and SUMOylation
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions