Learn More
Medchemexpress LLC (S)-2-Amino-6-((tert-butoxycarbonyl)amino)hexanoic acid | 2418-95-3 | 246.30 | 50 G
SDP
Catalog No. 5000448742
Encompass_Preferred
(S)-2-Amino-6-((tert-butoxycarbonyl)amino)hexanoic acid is a lysine derivative recognized for its role as an ergogenic dietary substance. It is utilized in commercial applications to influence anabolic hormone secretion, support fuel supply during exercise, enhance mental performance during stress-related tasks, and aid in preventing exercise-induced muscle damage.
  • High purity: 98.0%
  • White to off-white solid appearance
  • Soluble in H2O at 2 mg/mL (8.12 mM)
  • Influences anabolic hormone secretion
  • Supports fuel supply during exercise
  • Enhances mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
Catalog No. 50-004-48742 Supplier Medchemexpress LLC Supplier No. HYW00807950G
(S)-2-Amino-6-((tert-butoxycarbonyl)amino)hexanoic acid is a lysine derivative recognized for its role as an ergogenic dietary substance. It is utilized in commercial applications to influence anabolic hormone secretion, support fuel supply during exercise, enhance mental performance during stress-related tasks, and aid in preventing exercise-induced muscle damage.
  • High purity: 98.0%
  • White to off-white solid appearance
  • Soluble in H2O at 2 mg/mL (8.12 mM)
  • Influences anabolic hormone secretion
  • Supports fuel supply during exercise
  • Enhances mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage