Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-(2-aminoethyl) methanethiosulfonate hydrobromide | 16599-33-0 | 97.9% | 25 MG
SDP

Catalog No. 5000233534
Encompass_Preferred

MTSEA hydrobromide is a sulfhydryl-reactive reagent that selectively modifies free cysteine residues to introduce a positively charged side chain roughly the size of lysine. It is used in biochemical and electrophysiological studies to probe cysteine accessibility and to introduce charge at engineered cysteine sites.

  • Sulfhydryl-reactive reagent for targeted cysteine modification.
  • High supplied purity of 97.9%.
  • Molecular weight 236.15 g/mol; formula C3H10BrNO2S2.
  • Available in small solid packs suitable for research use.
  • Store sealed at -20°C; in solution store at -80°C for long-term stability.
  • CAS number 16599-33-0 for unambiguous identification.

Catalog No. 50-002-33534 Supplier Medchemexpress LLC Supplier No. HY12012825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MTSEA hydrobromide is a sulfhydryl-reactive reagent that selectively modifies free cysteine residues to introduce a positively charged side chain roughly the size of lysine. It is used in biochemical and electrophysiological studies to probe cysteine accessibility and to introduce charge at engineered cysteine sites.

  • Sulfhydryl-reactive reagent for targeted cysteine modification.
  • High supplied purity of 97.9%.
  • Molecular weight 236.15 g/mol; formula C3H10BrNO2S2.
  • Available in small solid packs suitable for research use.
  • Store sealed at -20°C; in solution store at -80°C for long-term stability.
  • CAS number 16599-33-0 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.