Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Aminoheptane, ChiPros 99%, ee 99+%, Thermo Scientific™

Catalog No. AAL1930714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19307-14 25 g
AAL19307-03 1 g
AAL19307-06 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL19307-14 Supplier Thermo Scientific Chemicals Supplier No. L1930714

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-(+)-2-Aminoheptane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water (5g/L at 20°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 44745-29-1
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00066324
InChI Key VSRBKQFNFZQRBM-ZETCQYMHSA-N
Synonym s-+-2-aminoheptane, 2-heptanamine, 2s, unii-1n3l0r99qb, s-2-aminoheptane, 2s-heptan-2-amine, tuaminoheptane, +, s-heptan-2-amine, s-heptane-2-amine, s-ahp
PubChem CID 2734517
IUPAC Name (2S)-heptan-2-amine
SMILES CCCCCC(C)N

Specifications

Density 0.766
Boiling Point 142°C to 144°C
Flash Point 54°C (129°F)
Refractive Index 1.419
Quantity 25 g
UN Number UN2733
Beilstein 1719086
Sensitivity Air Sensitive
Solubility Information Soluble in water (5g/L at 20°C)
Optical Rotation +7.2° (Neat)
Formula Weight 115.22
Percent Purity 99%
Assay ee ≥99%
Chemical Name or Material (S)-(+)-2-Aminoheptane
Show More Show Less
Hazard Category H226-H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 623-590-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.