Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Aminohexane, ChiPros 99+%, ee 99+%, Thermo Scientific™

Catalog No. AAL1930514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1930514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1930514 Supplier Thermo Scientific Chemicals Supplier No. L1930514

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-(+)-2-Aminohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water (17g/L at 20°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70492-67-0
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00671626
InChI Key WGBBUURBHXLGFM-UHFFFAOYNA-N
Synonym s-2-aminohexane, 2s-hexan-2-amine, s-2-hexylamine, s-+-2-aminohexane, s-+-2-aminohexane, chipros, ee 99+%, s-2-hexaneamine, pubchem6750, 2s-2-hexanamine, 2-hexanamine, 2s, +-s-2-aminohexane
PubChem CID 6999846
SMILES CCCCC(C)N

Specifications

Melting Point <-20°C
Density 0.755
Boiling Point 116°C
Flash Point −3°C (26°F)
Quantity 25 g
UN Number UN2733
Beilstein 5237601
Sensitivity Air Sensitive
Solubility Information Soluble in water (17g/L at 20°C)
IUPAC Name hexan-2-amine
Formula Weight 101.19
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(+)-2-Aminohexane
Show More Show Less
Hazard Category H225-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 622-789-6
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.