Learn More
Medchemexpress LLC (S)-2-Aminohexanedioic acid | 1118-90-7 | 98.2% | 161.16 | 250 MG
SDP
Catalog No. 5000366303
Encompass_Preferred
(S)-2-Aminohexanedioic acid (Standard) is an analytical standard used in research and analytical applications. It functions as a biochemical reagent and an organic compound for life science research, particularly as an analytical reference standard for qualitative, quantitative, and methodological research experiments in techniques such as HPLC, GC, and MS. This compound serves as a glutamine synthetase inhibitor, with a Ki value of 209 μM, and has been identified as a gliotoxin affecting astroglial cells.
  • Used as an analytical standard in research.
  • Intended for analytical applications.
  • Functions as a biochemical reagent for life science research.
  • Suitable for qualitative, quantitative, and methodological research experiments.
  • Supports analysis by HPLC, GC, and MS.
  • Acts as a glutamine synthetase inhibitor.
  • Identified as an endogenous metabolite.
  • Serves as a reference standard.
Catalog No. 50-003-66303 Supplier Medchemexpress LLC Supplier No. 250MGHYW013665R
Add to Cart
Add to Cart
(S)-2-Aminohexanedioic acid (Standard) is an analytical standard used in research and analytical applications. It functions as a biochemical reagent and an organic compound for life science research, particularly as an analytical reference standard for qualitative, quantitative, and methodological research experiments in techniques such as HPLC, GC, and MS. This compound serves as a glutamine synthetase inhibitor, with a Ki value of 209 μM, and has been identified as a gliotoxin affecting astroglial cells.
  • Used as an analytical standard in research.
  • Intended for analytical applications.
  • Functions as a biochemical reagent for life science research.
  • Suitable for qualitative, quantitative, and methodological research experiments.
  • Supports analysis by HPLC, GC, and MS.
  • Acts as a glutamine synthetase inhibitor.
  • Identified as an endogenous metabolite.
  • Serves as a reference standard.