Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-2-(Aminomethyl)Pyrrolidine

Catalog No. 422886500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-288-6500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 42-288-6500MG Supplier Sigma Aldrich Supplier No. 422886500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-2-(Aminomethyl)pyrrolidine is a chiral vicinal diamine.Used in the synthesis of compounds with antitumor activity.

Specifications

CAS 60456-22-6
Density 1.103 g/mL at 25°C
Boiling Point 63°C (37 mmHg, lit.)
Molecular Formula C6H11ClO2
Refractive Index n20/D 1.482
Linear Formula C6H11ClO2
MDL Number MFCD00273365
Quantity 500 mg
Molecular Weight (g/mol) 150.6 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.