Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-(Aminomethyl)pyrrolidine 95.0+%, TCI America™
SDP

Catalog No. A2206200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2206200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2206200MG Supplier TCI America Supplier No. A2206200MG

Chemical Identifiers

CAS 69500-64-7
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.17
MDL Number MFCD00191745
InChI Key AUKXFNABVHIUAC-UHFFFAOYNA-N
Synonym s-pyrrolidin-2-ylmethanamine, s-+-2-aminomethyl pyrrolidine, s-2-aminomethyl pyrrolidine, 2s-pyrrolidin-2-ylmethylamine, s-pyrrolidin-2-ylmethylamine, s-2-aminomethylpyrrolidine, 2s-pyrrolidin-2-yl methanamine, 2-pyrrolidinemethanamine, 2s, 1-2s-pyrrolidin-2-yl methanamine, d07vdj
PubChem CID 2734054
ChEBI CHEBI:44632
IUPAC Name 1-(pyrrolidin-2-yl)methanamine
SMILES NCC1CCCN1

Specifications

Color Colorless
Boiling Point 85°C
Quantity 200 mg
UN Number 2734
Formula Weight 100.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-2-(Aminomethyl)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.