Learn More
Medchemexpress LLC (S)-2-Aminopropanamide hydrochloride | 33208-99-0 | ≥97.0% | C3H9ClN2O | 25 G
SDP
Catalog No. 5000358566
Encompass_Preferred
(S)-2-Aminopropanamide hydrochloride is a biochemical reagent suitable for use as a biological material or organic compound in life science related research.
  • White to off-white solid appearance
  • For research use only
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
Catalog No. 50-003-58566 Supplier Medchemexpress LLC Supplier No. 25GHY75326
Add to Cart
Add to Cart
(S)-2-Aminopropanamide hydrochloride is a biochemical reagent suitable for use as a biological material or organic compound in life science related research.
  • White to off-white solid appearance
  • For research use only
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month