Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-2-Aminopropanamide (hydrochloride) | 33208-99-0 | ≥97.0% | C3H9ClN2O | 50 G
SDP

Catalog No. 5000434326
Encompass_Preferred

This chemical is (S)-2-Aminopropanamide (hydrochloride), identified by CAS Number 33208-99-0. It has a molecular formula of C3H9ClN2O and a purity of ≥97.0%. It is a white to off-white solid.

  • CAS Number: 33208-99-0
  • Purity: ≥97.0%
  • Molecular formula: C3H9ClN2O
  • Molecular weight: 124.57
  • Appearance: White to off-white solid
  • Storage conditions for powder: -20°C for 3 years; 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-34326 Supplier Medchemexpress LLC Supplier No. HY7532650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This chemical is (S)-2-Aminopropanamide (hydrochloride), identified by CAS Number 33208-99-0. It has a molecular formula of C3H9ClN2O and a purity of ≥97.0%. It is a white to off-white solid.

  • CAS Number: 33208-99-0
  • Purity: ≥97.0%
  • Molecular formula: C3H9ClN2O
  • Molecular weight: 124.57
  • Appearance: White to off-white solid
  • Storage conditions for powder: -20°C for 3 years; 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.