Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-(Anilinomethyl)pyrrolidine 98.0+%, TCI America™
SDP

Catalog No. A09455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09455G 5 g
A09451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09455G Supplier TCI America Supplier No. A09455G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64030-44-0
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
MDL Number MFCD00015890
InChI Key MCHWKJRTMPIHRA-NSHDSACASA-N
Synonym s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline
PubChem CID 6950385
IUPAC Name N-[[(2S)-pyrrolidin-2-yl]methyl]aniline
SMILES C1CC(NC1)CNC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 126°C
Quantity 5 g
UN Number 1760
Formula Weight 176.26
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-(Anilinomethyl)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.