Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Bromopropionic acid, 98%, >85% ee

Catalog No. AC277910250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC277910250 25 g Glass bottle
AC277910050 5 g Glass bottle
AC277911000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC277910250 Supplier Thermo Scientific Chemicals Supplier No. 277910250
Add to Cart

CAS: 32644-15-8 | C3H5BrO2 | 152.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32644-15-8
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00137325
InChI Key MONMFXREYOKQTI-REOHCLBHSA-N
Synonym s---2-bromopropionic acid, s-2-bromopropionic acid, 2s-2-bromopropanoic acid, s-2-bromopropanoic acid, l-2-bromopropionic acid, s---2-bromopropionicacid, l-alpha-bromopropionic acid, 2s---2-bromopropionic acid, s-alpha-bromopropionic acid
PubChem CID 642232
IUPAC Name (2S)-2-bromopropanoic acid
SMILES CC(C(=O)O)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -35°C
Color Colorless
Density 1.7000g/mL
Boiling Point 203°C
Flash Point 100°C
Enantiomeric Excess 80% min. (Typically)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.4740 to 1.476
Linear Formula CH3CH(Br)CO2H
Quantity 25 g
Beilstein 02, 254
Specific Gravity 1.7
Specific Rotation -27
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Formula Weight 152.98
Percent Purity 98%,>85% ee
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Bromopropionic acid, >85%ee
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.