Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-2-Butanol

Catalog No. 23769810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-769-810G 10 g
23-769-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 23-769-810G Supplier Sigma Aldrich Supplier No. 23769810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-2-Butanol can be used:To synthesize chiral asymmetric perylene diimides (PDI) that spontaneously self-assemble into one-handed nanotubes that are photoconductive or fluorescent in nature.To synthesize bulky chiral vinyl monomers that can be radically polymerized to helical polymers with an excess screw sense.As chiral solvent to study excited state photo transfer of the photoacid, 5,8-dicyano-2-naphthol (DCN2) in chiral environment.

Specifications

CAS 4221-99-2
Density 0.803 g/mL at 25°C
Boiling Point 99°C to 100°C
Molecular Formula C4H10O
Refractive Index n20/D 1.397
Linear Formula C2H5CH(OH)CH3
MDL Number MFCD00064281
Quantity 10 g
Synonym (S)-(+)-sec-Butyl alcohol
Molecular Weight (g/mol) 74.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.