Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-(2-carboxyethyl)-L-cysteine | 4033-46-9 | MFCD03424208 | 98.6% | 193.22 g·mol⁻1 | C6H11NO4S | 10 MG
SDP

Catalog No. 5000283017
Encompass_Preferred

S-(2-Carboxyethyl)-L-cysteine is an amino acid-derived research reagent provided as a solid for biochemical and pharmacological studies that require a cysteine derivative. The compound is characterized with analytical data to support research use and is offered in small quantities for laboratory experiments.

  • High purity: 98.6%.
  • Chemical formula: C6H11NO4S.
  • Molecular weight: 193.22 g·mol⁻1.
  • Physical form: solid; soluble in water at 5 mg/mL with ultrasonic assistance.
  • Available sizes: includes 10 mg and 25 mg; larger sizes available by quote.
  • Supporting documents: data sheet, COA, SDS, HNMR, RP-HPLC, MS.

Catalog No. 50-002-83017 Supplier Medchemexpress LLC Supplier No. HYW14177410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-(2-Carboxyethyl)-L-cysteine is an amino acid-derived research reagent provided as a solid for biochemical and pharmacological studies that require a cysteine derivative. The compound is characterized with analytical data to support research use and is offered in small quantities for laboratory experiments.

  • High purity: 98.6%.
  • Chemical formula: C6H11NO4S.
  • Molecular weight: 193.22 g·mol⁻1.
  • Physical form: solid; soluble in water at 5 mg/mL with ultrasonic assistance.
  • Available sizes: includes 10 mg and 25 mg; larger sizes available by quote.
  • Supporting documents: data sheet, COA, SDS, HNMR, RP-HPLC, MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.