Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-(2-Carboxyethyl)-L-cysteine 98.0+%, TCI America™
SDP

Catalog No. C2781100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2781100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2781100MG Supplier TCI America Supplier No. C2781100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4033-46-9
Molecular Formula C6H11NO4S
Molecular Weight (g/mol) 193.217
MDL Number MFCD03424208
InChI Key FBPINGSGHKXIQA-BYPYZUCNSA-N
Synonym 3-[(2-Carboxyethyl)thio]alanine
PubChem CID 23618202
IUPAC Name (2R)-2-amino-3-(2-carboxyethylsulfanyl)propanoic acid
SMILES C(CSCC(C(=O)O)N)C(=O)O

Specifications

Melting Point 198°C
Color White
Quantity 100 mg
Formula Weight 193.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material S-(2-Carboxyethyl)-L-cysteine
RTECSNumber AY4300000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.