Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-Chlorobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. C13771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13771G 1 g
C13775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13771G Supplier TCI America Supplier No. C13771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32653-32-0
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00067099
InChI Key RVBUZBPJAGZHSQ-VKHMYHEASA-N
PubChem CID 6999872
IUPAC Name (2S)-2-chlorobutanoic acid
SMILES CCC(C(=O)O)Cl

Specifications

Color Yellow
Boiling Point 94°C
Quantity 1 g
UN Number 3265
Formula Weight 122.55
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (S)-2-Chlorobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.