Learn More
Medchemexpress LLC (S)-2-(dimethylcarbamothioyl)-N-(2-methyl-4-(1-methyl-1,4,5,10-tetrahydrobenzo[b]pyrazolo | 2245072-21-1 | 98.6% | 645.70 g/mol | C30H34F3N7O4S | 10MG
SDP
Catalog No. 5000235545
Encompass_Preferred
LIT-001 is a nonpeptide oxytocin receptor (OT-R) agonist for preclinical research, reported to have EC50 = 55 nM and Ki = 226 nM at OT-R and to improve social interaction in a mouse model of autism. The compound is provided as a high-purity solid and as 10 mM solutions for in vitro and in vivo studies, with documented solubility and storage recommendations.
Catalog No. 50-002-35545 Supplier Medchemexpress LLC Supplier No. HY124733A10MG
LIT-001 is a nonpeptide oxytocin receptor (OT-R) agonist for preclinical research, reported to have EC50 = 55 nM and Ki = 226 nM at OT-R and to improve social interaction in a mouse model of autism. The compound is provided as a high-purity solid and as 10 mM solutions for in vitro and in vivo studies, with documented solubility and storage recommendations.