Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Heptanol 98%, Thermo Scientific™

Catalog No. AC290390010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC290390010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290390010 Supplier Thermo Scientific Chemicals Supplier No. 290390010

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6033-23-4
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
InChI Key CETWDUZRCINIHU-ZETCQYMHSA-N
Synonym s-+-2-heptanol, 2s-heptan-2-ol, 2-heptanol, s, unii-7xq9f7kypw, heptan-2s-ol, s-2-heptanol, s-heptan-2-ol, 7xq9f7kypw, d-2-heptanol
PubChem CID 2724897
ChEBI CHEBI:87378
IUPAC Name (2S)-heptan-2-ol
SMILES CCCCCC(C)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.81
Infrared Spectrum Authentic
Refractive Index 1.4200 to 1.4220 (20°C, 589nm)
Linear Formula CH3(CH2)4CH(OH)CH3
Quantity 1 g
Beilstein 01, 205
Merck Index 15, 4697
Specific Rotation +9° to +11° (20°C, 589nm) (neat)
Formula Weight 116.2
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Heptanol, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.