Learn More
Medchemexpress LLC (S)-2-hydroxypropanoic acid sodium | 867-56-1 | MFCD00066576 | 98.0% | 112.06 | C3H5NaO3 | 25 MG
SDP
Catalog No. 5000209076
Encompass_Preferred
L-lactic acid sodium is the sodium salt of L-lactic acid used as a chemical building block and research reagent. It can serve as a precursor for polylactic acid synthesis and has reported antiproliferative activity. For research use only; not for human or veterinary use.
  • Sodium salt of L-lactic acid, water-soluble.
  • Used as a precursor for polylactic acid and other chemical syntheses.
  • Reported antiproliferative activity in research studies.
  • Available in small milligram pack sizes suitable for laboratory use.
  • Typical purity: 98.0%.
Catalog No. 50-002-09076 Supplier Medchemexpress LLC Supplier No. 25MGHYW040233
L-lactic acid sodium is the sodium salt of L-lactic acid used as a chemical building block and research reagent. It can serve as a precursor for polylactic acid synthesis and has reported antiproliferative activity. For research use only; not for human or veterinary use.
  • Sodium salt of L-lactic acid, water-soluble.
  • Used as a precursor for polylactic acid and other chemical syntheses.
  • Reported antiproliferative activity in research studies.
  • Available in small milligram pack sizes suitable for laboratory use.
  • Typical purity: 98.0%.