Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific™

Catalog No. AC362102500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC362102500 250 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC362102500 Supplier Thermo Scientific Chemicals Supplier No. 362102500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213270-36-1
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD01091033
InChI Key AJNNFMRCMKGYHA-ZETCQYMHSA-N
Synonym s-3-methoxycarbonyl-5-methylhexanoic acid, s---2-isobutylsuccinic acid 1-methyl ester, 3s-3-methoxycarbonyl-5-methylhexanoic acid, s-2-isobutylsuccinic acid 1-methyl ester, s-2-2-methylpropyl butanedioic acid 1-methyl ester, butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2s
PubChem CID 2734513
IUPAC Name (3S)-3-methoxycarbonyl-5-methylhexanoic acid
SMILES CC(C)CC(CC(=O)O)C(=O)OC

Specifications

Packaging Glass Bottle
Quantity 250 mg
Formula Weight 188.22
Percent Purity 95%,(98% ee)
Chemical Name or Material (S)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.