Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-2-(Methoxydiphenylmethyl)Pyrrolidine

Catalog No. 670197500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-019-7500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 67-019-7500MG Supplier Sigma Aldrich Supplier No. 670197500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-2-(Methoxydiphenylmethyl)pyrrolidine is a diphenylprolinol methyl ether, which can be used as a catalyst to synthesize: Enantioselective ketones via intermolecular asymmetric Michael addition of aldehydes to nonactivated enones. Stereoselective chiral bipyrazolidin-3-one derivatives by dipolar cycloaddition reaction of azomethine imines with +a, +a-unsaturated aldehydes. Enantioenriched spiro nitrogen heterocycles via asymmetric nucleophilic epoxidation of +a-ylideneoxindole esters. Optically active secondary alcohols by asymmetric addition of Et2Zn to various aldehydes.

Specifications

CAS 132958-72-6
Density 0.899 g/mL at 25°C
Boiling Point 166°C (760 mmHg, lit.)
Molecular Formula C6H14N2
Linear Formula C6H14N2
Quantity 500 mg
MDL Number MFCD00191347
Molecular Weight (g/mol) 114.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.