Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Methyl-1-butanol 98.0+%, TCI America™
SDP

Catalog No. M017025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M017025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M017025ML Supplier TCI America Supplier No. M017025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1565-80-6
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00064299
InChI Key QPRQEDXDYOZYLA-YFKPBYRVSA-N
Synonym s---2-methyl-1-butanol, s-2-methyl-1-butanol, s-2-methylbutan-1-ol, 1-butanol, 2-methyl-, 2s, 2s-2-methylbutan-1-ol, unii-ii2qjb35ic, 1-butanol, 2-methyl-, s, 2s-2-methyl-1-butanol, s---2-methylbutanol, 2-methyl-1-butanol,-
PubChem CID 2723602
ChEBI CHEBI:50625
IUPAC Name (2S)-2-methylbutan-1-ol
SMILES CCC(C)CO

Specifications

Color Colorless
Boiling Point 128°C
Quantity 25 mL
UN Number 1105
Formula Weight 88.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Methyl-1-butanol
EINECSNumber (2)-0217
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.