Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-2-Methyl-Cbs-Oxazaborolidine Solution

Catalog No. 67464825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-464-825ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 67-464-825ML Supplier Sigma Aldrich Supplier No. 67464825ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The CBS (Corey-Bakshi-Shibata) oxazaborolidine catalyst has been used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of +a-hydroxy acids, +a-amino acids, C2-symmetrical ferrocenyl diols, and propargyl alcohols.

Specifications

CAS 161513-47-9
Melting Point 48°C to 50°C
Molecular Formula C8H15NO3
Refractive Index n20/D 1.460
Linear Formula C8H15NO3
MDL Number MFCD06200796
Quantity 1 g
Synonym tert-Butyl (2S)-N-(2-oxiranylmethyl)carbamate
Molecular Weight (g/mol) 173.21 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.