Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-2-Methyl-Cbs-Oxazaborolidine Solution

Catalog No. 45770125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-770-125ML 25 mL
45-770-15ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 45-770-125ML Supplier Sigma Aldrich Supplier No. 45770125ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The CBS (Corey-Bakshi-Shibata) oxazaborolidine catalyst has been used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of +a-hydroxy acids, +a-amino acids, C2-symmetrical ferrocenyl diols, and propargyl alcohols.Precipitate may form in storage, but does not affect product quality. Gently heat product to 80-90 C under an inert atmosphere to redissolve solids.

Specifications

CAS 112022-81-8
Density 0.929 g/mL at 25°C
Boiling Point 111°C
Molecular Formula C18H20BNO
Linear Formula C18H20BNO
MDL Number MFCD00078439
Quantity 25 mL
Synonym alpha,alpha-Diphenyl-L-prolinol methylboronic acid cycl-amide ester; (S)-1-Methyl-3,3-diphenyl-tetrahydro-pyrrolo[1,2c][1,3,2]oxazaborole; (S)-3,3-Diphenyl-1-methyltetrahydro-3H-pyrrolo[1,2-c][1,3,2]oxazaborole; (S)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole
Molecular Weight (g/mol) 277.17 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.