Learn More
Medchemexpress LLC (S)-2-(Methylamino)-3-phenylpropanoic acid | 2566-30-5 | 179.22 | 100 G
SDP
Catalog No. 5000445496
Encompass_Preferred
(S)-2-(Methylamino)-3-phenylpropanoic acid is a phenylalanine derivative. Amino acids and amino acid derivatives, such as this product, are used as ergogenic supplements due to their influence on anabolic hormone secretion, fuel supply during exercise, mental performance during stress-related tasks, and prevention of exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
  • Solid appearance
  • White to off-white color
  • Derived from microorganisms, Lasiosphaera Seu Calvatia
  • Soluble in H₂O at 40 mg/mL
  • For research use only
Catalog No. 50-004-45496 Supplier Medchemexpress LLC Supplier No. HYW005388100G
(S)-2-(Methylamino)-3-phenylpropanoic acid is a phenylalanine derivative. Amino acids and amino acid derivatives, such as this product, are used as ergogenic supplements due to their influence on anabolic hormone secretion, fuel supply during exercise, mental performance during stress-related tasks, and prevention of exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
  • Solid appearance
  • White to off-white color
  • Derived from microorganisms, Lasiosphaera Seu Calvatia
  • Soluble in H₂O at 40 mg/mL
  • For research use only