Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-2-Methylbutanol

Catalog No. A8340710G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A8340710G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. A8340710G Supplier Sigma Aldrich Supplier No. A8340710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-2-Methylbutanol can be used to prepare:(S)-(-)-2-Methyl-1-butyloxy carbonyl amino hexyl isocyanate (MBI), which is used to synthesize isocyanate copolymers.Chiral alkoxynaphathoic acid derivatives having liquid crystalline properties.3,4-Bis[(S)-2-methylbutoxy]thiophene, a key intermediate for the synthesis of polythiophenes.

Specifications

CAS 27894-50-4
Molecular Formula C15H22N2O4 -+ HCl
Refractive Index n/D 1.41
Linear Formula C6H5CH2OOCNH(CH2)4CH(NH2)COOCH3 -+ HCl
MDL Number MFCD00034846
Quantity 5 g
Synonym (S)-(-)-2-Methyl-1-butanol; Active amyl alcohol
Molecular Weight (g/mol) 88.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.