Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-2-Methylbutanol

Catalog No. 65980500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-980-500ML 500 mL
65-980-100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 65-980-500ML Supplier Sigma Aldrich Supplier No. 65980500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

rest mainly 3-methyl-1-butanol(S)-(-)-2-Methylbutanol can be used as a reactant to prepare:Methylbutyl-2-(3-thienyl)acetate (MBTA) by esterification reaction with 3-thiophene acetic acid in the presence of an acid catalyst.(2S)-2-Methyl-1-butanesulfenyl chloride by reacting with thioacetic acid using Mitsunobu reaction conditions.(+)-Violapyrone C and (-)-myxalamide A.

Specifications

CAS 1565-80-6
Density 0.819 g/mL at 20°C, 0.811 g/mL at 25°C
Boiling Point 129°C to 131°C
Molecular Formula C5H12O
Refractive Index n/D 1.41; n20/D 1.411
Linear Formula C5H12O
MDL Number MFCD00064299
Quantity 500 mL
Synonym (S)-(-)-2-Methyl-1-butanol; Active amyl alcohol
Molecular Weight (g/mol) 88.15 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.