Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-2-Methylbutyric Acid

Catalog No. 2455265G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-552-65G 5 g
24-552-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 24-552-65G Supplier Sigma Aldrich Supplier No. 2455265G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-2-Methylbutyric acid may be used in the preparation of the stereoisomers of ™-lavandulyl (S)-2-methylbutanoate and ™-maconelliyl (S)-2-methylbutanoate, which are female sex pheromones found in pink hibiscus mealybug.

Specifications

CAS 1730-91-2
Density 0.938 g/mL at 25°C
Boiling Point 78°C (15 mmHg, lit.)
Molecular Formula C5H10O2
Linear Formula C2H5CH(CH3)CO2H
Refractive Index n20/D 1.406
Quantity 5 g
MDL Number MFCD00063166
Molecular Weight (g/mol) 102.13 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.