Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Methylpiperazine, 99+%, ee 99+%, Thermo Scientific™

Catalog No. AAL2007503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2007503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2007503 Supplier Thermo Scientific Chemicals Supplier No. L2007503

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 74879-18-8
Melting Point 89°C to 91°C
Boiling Point 155°C to 156°C
Flash Point 65°C (149°F)
Molecular Formula C5H12N2
Quantity 1 g
MDL Number MFCD00171452
UN Number UN2925
Beilstein 3647995
Sensitivity Air sensitive; Hygroscopic
Synonym s-+-2-methylpiperazine, s-2-methylpiperazine, 2s-2-methylpiperazine, piperazine, 2-methyl-, 2s, s-2-methyl-piperazine, unii-c5vi3s1yc0, 2-methylpiperazine, s, 2-s-methylpiperazine, s-mp, c5vi3s1yc0
InChI Key JOMNTHCQHJPVAZ-YFKPBYRVSA-N
SMILES CC1CNCCN1
IUPAC Name (2S)-2-methylpiperazine
Molecular Weight (g/mol) 100.165
PubChem CID 2734219
Formula Weight 100.17
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(+)-2-Methylpiperazine
Show More Show Less
Hazard Category H228-H314
Hazard Statement GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.