Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-2-Methylpiperidine

Catalog No. 5229021G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-290-21G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 52-290-21G Supplier Sigma Aldrich Supplier No. 5229021G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-2-Methylpiperidine has been used in the preparation of (S)-N-(3-(2-pipecolin-1-yl)propyl)phthalimide by reacting with N-(3-bromopropyl)phthalimide. It may also be used as a starting material in the multi-step synthesis of (2S,6S)-(+)-solenopsin A.

Specifications

CAS 26250-84-0
Melting Point 118°C to 121°C
Molecular Formula C7H9NOS
Refractive Index n20/D 1.446
Linear Formula CH3C6H4S(O)NH2
MDL Number MFCD06858375
Quantity 5 g
Molecular Weight (g/mol) 155.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.