Learn More
AdipoGen S-2-NMA
SDP
Catalog No. 502560475
Encompass_Preferred
Chemical. CAS 157134-51-5. Formula C13H12O3. MW 216.23. Synthetic Excellent chiral anisotropic NMR reagent. Used for the determination of the absolute configuration of secondary alcohols.
Catalog No. 50-256-0475 Supplier AdipoGen Supplier No. CDXM0213M500
Chemical. CAS 157134-51-5. Formula C13H12O3. MW 216.23. Synthetic Excellent chiral anisotropic NMR reagent. Used for the determination of the absolute configuration of secondary alcohols.