Learn More
Medchemexpress LLC (S)-(+)-2-phenylglycine methyl ester hydrochloride | 15028-39-4 | MFCD00077158 | 99.9% | 201.65 g/mol | C9H12ClNO2 | 100 G
SDP
Catalog No. 5000300975
Encompass_Preferred
(S)-(+)-2-Phenylglycine methyl ester hydrochloride is the S-enantiomer of 2-phenylglycine methyl ester presented as the hydrochloride salt. It is a chiral amino acid derivative used as a building block and intermediate in research chemistry, asymmetric synthesis, and preparation of optically active compounds. Supplied as a solid with documented purity and enantiomeric excess for research use.
  • High enantiomeric purity (100.0% ee) supporting stereospecific synthesis.
  • High chemical purity (99.9% by HPLC) for reliable analytical and synthetic use.
  • Molecular formula C9H12ClNO2 and molecular weight 201.65 g/mol.
  • White to off-white solid, convenient for handling and storage.
  • Stable under sealed refrigeration; COA and SDS available for traceability.
Catalog No. 50-003-00975 Supplier Medchemexpress LLC Supplier No. HYW019714100G
(S)-(+)-2-Phenylglycine methyl ester hydrochloride is the S-enantiomer of 2-phenylglycine methyl ester presented as the hydrochloride salt. It is a chiral amino acid derivative used as a building block and intermediate in research chemistry, asymmetric synthesis, and preparation of optically active compounds. Supplied as a solid with documented purity and enantiomeric excess for research use.
  • High enantiomeric purity (100.0% ee) supporting stereospecific synthesis.
  • High chemical purity (99.9% by HPLC) for reliable analytical and synthetic use.
  • Molecular formula C9H12ClNO2 and molecular weight 201.65 g/mol.
  • White to off-white solid, convenient for handling and storage.
  • Stable under sealed refrigeration; COA and SDS available for traceability.