Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-(Trifluoromethyl)piperidine, 97%

Catalog No. p-4380114
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446650010 1 g
AC446650050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446650010 Supplier Thermo Scientific Chemicals Supplier No. 446650010
Only null left
Add to Cart

CAS: 154727-51-2 | C6H10F3N | 153.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154727-51-2
Molecular Formula C6H10F3N
Molecular Weight (g/mol) 153.15
InChI Key NAXDEFXCCITWEU-YFKPBYRVSA-N
Synonym s-2-trifluoromethyl piperidine, 2s-2-trifluoromethyl piperidine, s-2-trifluoromethylpiperidine, 2alpha-trifluoromethyl piperidine
PubChem CID 42580051
IUPAC Name (2S)-2-(trifluoromethyl)piperidine
SMILES C1CCNC(C1)C(F)(F)F

Specifications

Boiling Point 80.0°C (200.0 Torr)
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 153.15
Percent Purity 97%
Chemical Name or Material (S)-2-(Trifluoromethyl)piperidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.