Learn More
CELLINK LLC S.200 PLATE 10XS.200 PL 10PK
SDP
Catalog No. NC2061764
Encompass
S.200 Plate - 10 Pack10 x S.200 Plate, including 96 I.DOT wells per plate. Effective ReservoirVolume: 80 µl, orifice diameter: 200 µm. SBS compatible, polypropylene.
Catalog No. NC2061764 Supplier CELLINK LLC Supplier No. D16110027287
Add to Cart
Add to Cart
S.200 Plate - 10 Pack10 x S.200 Plate, including 96 I.DOT wells per plate. Effective ReservoirVolume: 80 µl, orifice diameter: 200 µm. SBS compatible, polypropylene.