Learn More
MSC Devcon 20845 25 mL Cartridge Two Part Epoxy 4 to 5 min Working Time
Catalog No. 501113845
Encompass
Devcon 20845 25 mL Cartridge Two Part Epoxy 4 to 5 min Working Time
Catalog No. 50-111-3845 Supplier MSC Supplier No. 00261164
Devcon 20845 25 mL Cartridge Two Part Epoxy 4 to 5 min Working Time