Learn More
SCP Science Standard S-21+K, 500 ppm, 200 g
Catalog No. NC9785574
Encompass
Oil based metal calibration standards for use with ICP, AA, rotrode, XRF, flame emission and other instruments. S-21: Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn and K.
Catalog No. NC9785574 Supplier SCP Science Supplier No. 150021059
Add to Cart
Add to Cart
Oil based metal calibration standards for use with ICP, AA, rotrode, XRF, flame emission and other instruments. S-21: Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn and K.