Learn More
Diapharma Group, Inc. S-2238 CHROMOGENIX SUB 25MG
SDP
Catalog No. NC9550144
Encompass
Chromogenic substrate for thrombin Package: 25 mg Molecular wt: 625.6 Formula: H-D-Phe-Pip-Arg-pNA -- 2HCl Solubility: >10mmol/L in H 2 O
Catalog No. NC9550144 Supplier Diapharma Group, Inc. Supplier No. S820324
Chromogenic substrate for thrombin Package: 25 mg Molecular wt: 625.6 Formula: H-D-Phe-Pip-Arg-pNA -- 2HCl Solubility: >10mmol/L in H 2 O