Learn More
Medchemexpress LLC HY-112257 5mg Medchemexpress, S-23 CAS:1010396-29-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1122575MG
Encompass_Preferred
Medchemexpress, HY-112257 5mg S-23 CAS:1010396-29-8 S-23 is an orally active selective androgen receptor modulator (SARM) with a Ki of 1.7 nM. S-23 induces androgen receptor (AR)-mediated transcriptional activation in CV-1 cells. S-23 increases prostate, seminal vesicle, and levator ani muscle weights in castrated rats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0813