Learn More
Diapharma Group, Inc. Chromogenix S-2302, 25 mg
SDP
Catalog No. NC0267128
Encompass
Chromogenix S-2302 is a chromogenic substrate for plasma kallikrein.
Catalog No. NC0267128 Supplier Diapharma Group, Inc. Supplier No. S820340
Chromogenix S-2302 is a chromogenic substrate for plasma kallikrein.