Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-[1-(4-Methoxybenzenesulfonyl)-(4S,5S)-4,5-diphenyl-4,5-dihydro-1H-imidazol-2-yl]-1,1'-binaphthalene-2,2- 96+%, TCI America™
SDP

Catalog No. M276550MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M276550MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M276550MG Supplier TCI America Supplier No. M276550MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1621994-95-3
Molecular Formula C42H31N2O7PS
Molecular Weight (g/mol) 738.751
InChI Key IOZGNSVHJJRNOD-ZAQUEYBZSA-N
PubChem CID 102334043
SMILES COC1=CC=C(C=C1)S(=O)(=O)N2C(C(N=C2C3=CC4=CC=CC=C4C5=C3OP(=O)(OC6=C5C7=CC=CC=C7C=C6)O)C8=CC=CC=C8)C9=CC=CC=C9

Specifications

Melting Point 205°C
Color Yellow
Quantity 50 mg
Formula Weight 738.75
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3-[1-(4-Methoxybenzenesulfonyl)-(4S,5S)-4,5-diphenyl-4,5-dihydro-1H-imidazol-2-yl]-1,1′-bi
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.