Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-3,3'-Bis(2,4,6-Triisopropylphenyl)-1,1'-Binaphthyl-2,2'-Diyl Hydrogenphosphate

Catalog No. 689785500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-978-5500MG 500 mg
68-978-5100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 68-978-5500MG Supplier Sigma Aldrich Supplier No. 689785500MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.Versatile organocatalyst. Uses include asymmetric reductive amination, enantioselective Diels-Alder reactions and asymmetric counteranion-directed catalysis (ACDC). Can be used in cooperation with chiral iridium complexes to catalyze greener asymmetric reductive amination using hydrogen gas.Metal-Br++nsted Acid Cooperative Catalysis for Asymmetric Reductive Amination

Specifications

CAS 874948-63-7
Molecular Formula C50H57O4P
Linear Formula C50H57O4P
MDL Number MFCD10567009
Quantity 500 mg
Synonym (11bS)-4-Hydroxy-2,6-bis[2,4,6-tris(1-methylethyl)phenyl]dinaphtho[2,1-d:1',2'-f]-1,3,2-dioxaphosphepin 4-oxide; (S)-3,3'-Bis(2,4,6-triisopropylphenyl)-1,1'-bi-2-naphthol cyclic monophosphate; (S)-TRIP
Molecular Weight (g/mol) 752.96 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.