Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 99%

Catalog No. AAL13334ME
Change view
Click to view available options
Quantity:
500 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13334ME 500 mg
AAL1333404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13334ME Supplier Thermo Scientific Chemicals Supplier No. L13334ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 33878-99-8 | C11H14O2 | 178.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33878-99-8
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00213797
InChI Key DNHDRUMZDHWHKG-NSHDSACASA-N
Synonym s-+-3,4,8,8a-tetrahydro-8a-methyl-1,6 2h,7h-naphthalenedione, 8as-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-3,4,7,8-tetrahydro-2h-naphthalene-1,6-dione, s-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-1,2,3,4,6,7,8,8a-octahydronaphthalene-1,6-dione, 8as-+-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl-, 8as, wieland-mieschler ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione #
PubChem CID 736069
IUPAC Name (8aS)-8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione
SMILES CC12CCC(=O)C=C1CCCC2=O

Specifications

Melting Point 48°C to 52°C
Quantity 500 mg
Beilstein 1569979
Optical Rotation +100° (c=1 in Benzene)
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material (S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.