Learn More
Cayman Chemical S-3 5-DHPG 10mg
SDP
Catalog No. 502971716
Encompass_Preferred
An agonist of the group mGluR, binding both mGluR1a and mGluR5a (Ki = 0.9 and 3.9 µM, respectively) but not ionotropic glutamate receptors
Catalog No. 50-297-1716 Supplier Cayman Chemical Supplier No. 1441110 MG
An agonist of the group mGluR, binding both mGluR1a and mGluR5a (Ki = 0.9 and 3.9 µM, respectively) but not ionotropic glutamate receptors