Learn More
Strem, An Ascensus Company CAS# 380230-02-4. 100mg. (S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)bis[(1S)-1-phenylethyl]amine, min. 95%. MFCD04117688
SDP
Supplier:  Strem, An Ascensus Company 151521100MG
Encompass
CAS# 380230-02-4. 100mg. (S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)bis[(1S)-1-phenylethyl]amine, min. 95%. MFCD04117688. Molecular Weight: 539.60. Molecular Formula: C36H30NO2P. Color/form: off-white pwdr. Strem# 15-1521. http://www.strem.com/catalog/v/15-1521/
Catalog No. 50-901-10154