Learn More
Strem, An Ascensus Company CAS# 415918-91-1. 100mg. (S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)bis[(1R)-1-phenylethyl]amine, dichloromethane adduct, min. 95%. MFCD04117687
SDP
Catalog No. 5090110152
Encompass
CAS# 415918-91-1. 100mg. (S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)bis[(1R)-1-phenylethyl]amine, dichloromethane adduct, min. 95%. MFCD04117687. Molecular Weight: 539.60. Molecular Formula: C36H30NO2P. Color/form: white pwdr. Strem# 15-1520. http://www.strem.com/catalog/v/15-1520/
Catalog No. 50-901-10152 Supplier Strem, An Ascensus Company Supplier No. 151520100MG
CAS# 415918-91-1. 100mg. (S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)bis[(1R)-1-phenylethyl]amine, dichloromethane adduct, min. 95%. MFCD04117687. Molecular Weight: 539.60. Molecular Formula: C36H30NO2P. Color/form: white pwdr. Strem# 15-1520. http://www.strem.com/catalog/v/15-1520/