Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid, 95%

Catalog No. AAH63516MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63516MD 250 mg
AAH6351603 1 g
AAH6351606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63516MD Supplier Thermo Scientific Chemicals Supplier No. H63516MD

missing translation for 'validMainframeMsg'

CAS: 188970-92-5 | C22H22N2O6 | 410.426 g/mol

(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188970-92-5
Molecular Formula C22H22N2O6
Molecular Weight (g/mol) 410.426
MDL Number MFCD00273468
InChI Key MPVGCCAXXFLGIU-IBGZPJMESA-N
Synonym fmoc-dap alloc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-allyloxy carbonyl amino propanoic acid, fmoc-dap aloc-oh, fmoc-3-allyloxy carbonyl amino-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-prop-2-en-1-yloxy carbonyl amino propanoic acid, ambotzfaa1366, fmoc-3-allyloxycarbonylamino-l-ala-oh, fmoc-dap alloc-oh hplc, n-, a-fmoc-n-, a-allyloxycarbonyl-l-2,3-diaminopropion, na-fmoc-nb-allyloxycarbonyl-l-2,3-diaminopropionic acid
PubChem CID 2756107
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(prop-2-enoxycarbonylamino)propanoic acid
SMILES C=CCOC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 250 mg
Beilstein 9161065
Formula Weight 410.42
Percent Purity 95%
Physical Form Solid
Chemical Name or Material (S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.