Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-Amino-1-carbobenzoxypyrrolidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A23241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23241G 1 g
A23245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23241G Supplier TCI America Supplier No. A23241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 550378-39-7
Molecular Formula C12H17ClN2O2
Molecular Weight (g/mol) 256.73
MDL Number MFCD06796559
InChI Key QNQVBYGRFHOBNO-MERQFXBCSA-N
Synonym (S)-3-Amino-1-Cbz-pyrrolidine Hydrochloride
PubChem CID 20820180
IUPAC Name benzyl (3S)-3-aminopyrrolidine-1-carboxylate;hydrochloride
SMILES C1CN(CC1N)C(=O)OCC2=CC=CC=C2.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 256.73
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3-Amino-1-carbobenzoxypyrrolidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.